Skip to Content
Merck
vibrant-m

di-Potassium hydrogen phosphate anhydrous

vibrant-m
Synonym(s):
di-Potassium hydrogen phosphate trihydrate
Empirical Formula (Hill Notation):
HK2O4P · 3H2O
CAS Number:
Molecular Weight:
228.22

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing