Skip to Content
Merck
Dimethyl azelate analytical standard

Dimethyl azelate

Dimethyl azelate analytical standard
Synonym(s):
Dimethyl nonanedioate, Methyl azelate, Nonanedioic acid dimethyl ester
Linear Formula:
CH3OCO(CH2)7COOCH3
CAS Number:
Molecular Weight:
216.27

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing