![Dimethyl adipate ≥99%](/deepweb/assets/sigmaaldrich/product/structures/400/109/b94f6849-86e1-4320-ae22-cf840e2a337e/640/b94f6849-86e1-4320-ae22-cf840e2a337e.png)
Dimethyl adipate
![Dimethyl adipate ≥99%](/deepweb/assets/sigmaaldrich/product/structures/400/109/b94f6849-86e1-4320-ae22-cf840e2a337e/640/b94f6849-86e1-4320-ae22-cf840e2a337e.png)
Synonym(s):
Dimethyl adipate, Dimethyladipate
Linear Formula:
CH3OCO(CH2)4COOCH3
CAS Number:
Molecular Weight:
174.19
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing