Skip to Content
Merck
Dihydrocortisol ≥95% (HPLC)

Dihydrocortisol

Dihydrocortisol ≥95% (HPLC)
Synonym(s):
5β-Pregnane-11β,17α,21-triol-3,20-dione, 11β,17α,21-Trihydroxy-5β-pregnane-3,20-dione, 5β-Dihydrocortisol
Empirical Formula (Hill Notation):
C21H32O5
CAS Number:
Molecular Weight:
364.48

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing