![Didanosine Related Compound B United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/333/895/a4a92f0c-3f77-4d9b-a9c6-a34153c2956a/640/a4a92f0c-3f77-4d9b-a9c6-a34153c2956a.png)
Didanosine Related Compound B
![Didanosine Related Compound B United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/333/895/a4a92f0c-3f77-4d9b-a9c6-a34153c2956a/640/a4a92f0c-3f77-4d9b-a9c6-a34153c2956a.png)
Synonym(s):
2′,3′-Dideoxyadenosine, ddA
Empirical Formula (Hill Notation):
C10H13N5O2
CAS Number:
Molecular Weight:
235.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing