![Dicyclomine Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/157/655/543bfe59-a86f-4680-abd5-1fc79bb99152/640/543bfe59-a86f-4680-abd5-1fc79bb99152.jpg)
Dicyclomine Related Compound A
![Dicyclomine Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/157/655/543bfe59-a86f-4680-abd5-1fc79bb99152/640/543bfe59-a86f-4680-abd5-1fc79bb99152.jpg)
Synonym(s):
[1,1′-Bicyclohexyl]-1-carboxylic acid
Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing