Skip to Content
Merck
vibrant-m

Dichlorodifluoromethane solution

vibrant-m
Empirical Formula (Hill Notation):
CCl2F2
CAS Number:
Molecular Weight:
120.91

Products

Brand
Formula Weight
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1