Skip to Content
Merck
Decanal analytical standard

Decanal

Decanal analytical standard
Synonym(s):
Caprinaldehyde, Decyl aldehyde
Linear Formula:
CH3(CH2)8CHO
CAS Number:
Molecular Weight:
156.27
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing