Skip to Content
Merck
Cholecalciferol (Vitamin D3) Pharmaceutical Secondary Standard; Certified Reference Material

Cholecalciferol (Vitamin D3)

Cholecalciferol (Vitamin D3) Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Cholecalciferol, (+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1