
Carmine (C.I. 75470)

Synonym(s):
Carmine (C.I. 75470)
Empirical Formula (Hill Notation):
C44H37AlCaO27 · 3H2O
CAS Number:
Molecular Weight:
1118.85
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing