
Boc-Tyr(Bzl)-OH

Synonym(s):
Boc-Tyr(Bzl)-OH
Empirical Formula (Hill Notation):
C21H25NO5
CAS Number:
Molecular Weight:
371.43
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1