
Boc-Gly-OH-2-13C

Synonym(s):
N-(tert-Butoxycarbonyl)glycine-2-13C, Glycine-2-13C, N-t-Boc derivative
Linear Formula:
Boc-NH13CH2CO2H
CAS Number:
Molecular Weight:
176.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing