Skip to Content
Merck
Bicyclo[2.2.1]hepta-2,5-diene 98%

Bicyclo[2.2.1]hepta-2,5-diene

Bicyclo[2.2.1]hepta-2,5-diene 98%
Synonym(s):
2,5-Norbornadiene, NBD
Empirical Formula (Hill Notation):
C7H8
CAS Number:
Molecular Weight:
92.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing