Benodanil
Synonym(s):
2-Iodobenzanilide
Empirical Formula (Hill Notation):
C13H10INO
CAS Number:
Molecular Weight:
323.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing