![Ascorbyl palmitate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/138/578/86b241db-0800-438a-9383-9fb666f6de74/640/86b241db-0800-438a-9383-9fb666f6de74.png)
Ascorbyl palmitate
![Ascorbyl palmitate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/138/578/86b241db-0800-438a-9383-9fb666f6de74/640/86b241db-0800-438a-9383-9fb666f6de74.png)
Synonym(s):
6-O-Palmitoyl-L-ascorbic acid, L-Ascorbic acid 6-hexadecanoate, L-Ascorbyl palmitate, Ascorbic acid 6-palmitate, NSC 402451, L-Ascorbic acid 6-palmitate, 6-O-Palmitoyl-L-ascorbic acid
Empirical Formula (Hill Notation):
C22H38O7
CAS Number:
Molecular Weight:
414.53
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique