Articaine EP Impurity C
Synonym(s):
1-Methylethyl 4-methyl-3-[[(2RS)-2-(propylamino)-propanoyl]amino]thiophene-2-carboxylate hydrochloride, Articaine isopropyl ester hydrochloride
Empirical Formula (Hill Notation):
C15H24N2O3S · HCl
CAS Number:
Molecular Weight:
348.89
Products
Brand
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing