Skip to Content
Merck
Anti-TRPV4 antibody produced in rabbit affinity isolated antibody

Anti-TRPV4 antibody produced in rabbit

Anti-TRPV4 antibody produced in rabbit affinity isolated antibody
Synonym(s):
Anti-OTRPC4, Anti-Transient Receptor Potential Channel V4, Anti-TRP12, Anti-VR-OAC, Anti-VRL2

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique