![ANTI-SUN1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/414/730/541157ff-b165-4b9c-9c56-20bca91df584/640/541157ff-b165-4b9c-9c56-20bca91df584.jpg)
ANTI-SUN1 antibody produced in rabbit
![ANTI-SUN1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/414/730/541157ff-b165-4b9c-9c56-20bca91df584/640/541157ff-b165-4b9c-9c56-20bca91df584.jpg)
Synonym(s):
Anti-Sad1/unc-84 protein-like 1, Anti-UNC84A, Anti-Unc-84 homolog A
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing