Anti-sox2 antibody produced in rabbit
Synonym(s):
cb236, sox2, wu:fb83g04, wu:fc14d07, Anti-ANOP3, Anti-MGC2413, Anti-SRY (sex determining region Y)-box 2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-2 of 2