![Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat affinity isolated antibody, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/832/026/3711e71c-cb00-4c68-9b14-50c6fae70d28/640/3711e71c-cb00-4c68-9b14-50c6fae70d28.jpg)
Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat
![Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat affinity isolated antibody, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/832/026/3711e71c-cb00-4c68-9b14-50c6fae70d28/640/3711e71c-cb00-4c68-9b14-50c6fae70d28.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Recombinant Host
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing