Skip to Content
Merck
Antimony(III) oxide powder, 5 μm, ReagentPlus®, 99%

Antimony(III) oxide

Antimony(III) oxide powder, 5 μm, ReagentPlus®, 99%
Synonym(s):
Diantimony trioxide, Antimony(III) oxide
Linear Formula:
Sb2O3
CAS Number:
Molecular Weight:
291.52

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core