![Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal clone GT-34, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/156/492/8076f4a1-a65f-4ca3-a6ba-91f692f04ae3/640/8076f4a1-a65f-4ca3-a6ba-91f692f04ae3.jpg)
Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal
![Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal clone GT-34, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/156/492/8076f4a1-a65f-4ca3-a6ba-91f692f04ae3/640/8076f4a1-a65f-4ca3-a6ba-91f692f04ae3.jpg)
Synonym(s):
Monoclonal Anti-Goat/Sheep IgG
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing