
Anti-CD45RA Antibody (human), PerCP-Cy5.5, clone HI100

Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2