Skip to Content
Merck
vibrant-m

Almotriptan Related Compound C

vibrant-m
Synonym(s):
N-Methyl-2-{5-[(pyrrolidin-1-ylsulfonyl)methyl]-1H-indol-3-yl}ethanamine
Empirical Formula (Hill Notation):
C16H23N3O2S
CAS Number:
Molecular Weight:
321.44

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1