Skip to Content
Merck
7H-Dibenzo[c,g]carbazole BCR®, certified reference material

7H-Dibenzo[c,g]carbazole

7H-Dibenzo[c,g]carbazole BCR®, certified reference material
Synonym(s):
3,4,5,6-Dibenzocarbazole
Empirical Formula (Hill Notation):
C20H13N
CAS Number:
Molecular Weight:
267.32

Products

Formula Weight
Manufacturer
Agency
Format
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing