Skip to Content
Merck
4-Methylbenzhydrylamine resin hydrochloride 50-200 mesh

4-Methylbenzhydrylamine resin hydrochloride

4-Methylbenzhydrylamine resin hydrochloride 50-200 mesh
Synonym(s):
(α-Amino-α-p-xylyl hydrochloride)polystyrene crosslinked with divinylbenzene, Amino-(4-methylphenyl)methyl copolystyrene divinylbenzene, MBHA-polystyrene

Products

Product Category
Brand
Physical Form
Showing 1-2 of 2