Skip to Content
Merck
4-Hydroxybutanoic acid lactone ≥98%, FCC, FG

4-Hydroxybutanoic acid lactone

4-Hydroxybutanoic acid lactone ≥98%, FCC, FG
Synonym(s):
γ-Butyrolactone, γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL
Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing