Skip to Content
Merck
4,5-Dicyanoimidazole solution 0.25 M in acetonitrile

4,5-Dicyanoimidazole solution

4,5-Dicyanoimidazole solution 0.25 M in acetonitrile
Synonym(s):
Imidazole-4,5-dicarbonitrile solution
Empirical Formula (Hill Notation):
C5H2N4
CAS Number:
Molecular Weight:
118.10

Products

Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing