![4,5,6,7-Tetrahydroindole 98%](/deepweb/assets/sigmaaldrich/product/structures/262/111/a6380210-8978-4cf4-b927-1f4b8eb3d18b/640/a6380210-8978-4cf4-b927-1f4b8eb3d18b.png)
4,5,6,7-Tetrahydroindole
![4,5,6,7-Tetrahydroindole 98%](/deepweb/assets/sigmaaldrich/product/structures/262/111/a6380210-8978-4cf4-b927-1f4b8eb3d18b/640/a6380210-8978-4cf4-b927-1f4b8eb3d18b.png)
Synonym(s):
2,3-Tetramethylenepyrrole, Cyclohex[b]pyrrole, NSC 122455
Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing