Skip to Content
Merck
3-m-Tolyl-1H-pyrazol-5-amine AldrichCPR

3-m-Tolyl-1H-pyrazol-5-amine

3-m-Tolyl-1H-pyrazol-5-amine AldrichCPR
Empirical Formula (Hill Notation):
C10H11N3
CAS Number:
Molecular Weight:
173.21

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing