
3-Mercapto-3-methyl-1-butyl-1-formate

Empirical Formula (Hill Notation):
C6H12O2S
CAS Number:
Molecular Weight:
148.22
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing