![3,3′-Diindolylmethane analytical standard](/deepweb/assets/sigmaaldrich/product/structures/195/700/ce5d75fd-10dc-4eb5-8027-a842030100b3/640/ce5d75fd-10dc-4eb5-8027-a842030100b3.png)
3,3′-Diindolylmethane
![3,3′-Diindolylmethane analytical standard](/deepweb/assets/sigmaaldrich/product/structures/195/700/ce5d75fd-10dc-4eb5-8027-a842030100b3/640/ce5d75fd-10dc-4eb5-8027-a842030100b3.png)
Synonym(s):
3,3′-Bisindolylmethane, DIM
Empirical Formula (Hill Notation):
C17H14N2
CAS Number:
Molecular Weight:
246.31
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing