2-Isopropyl-3-methoxypyrazine
Empirical Formula (Hill Notation):
C8H12N2O
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing