Skip to Content
Merck
2-(Diisopropylamino)-ethylchloride hydrochloride for synthesis

2-(Diisopropylamino)-ethylchloride hydrochloride

2-(Diisopropylamino)-ethylchloride hydrochloride for synthesis
Synonym(s):
2-(Diisopropylamino)-ethylchloride hydrochloride
Empirical Formula (Hill Notation):
C8H19Cl2N
CAS Number:
Molecular Weight:
200.15

Products

Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing