Skip to Content
Merck
2-Amino-2-methyl-1-propanol BioUltra, ≥99.0% (GC)

2-Amino-2-methyl-1-propanol

2-Amino-2-methyl-1-propanol BioUltra, ≥99.0% (GC)
Synonym(s):
β-Aminoisobutyl alcohol, AMP, 2-Amino-2-methyl-1-propanol
Linear Formula:
(CH3)2C(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade