Skip to Content
Merck
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥99%

2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole

2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥99%
Synonym(s):
2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole, Butyl-PBD
Empirical Formula (Hill Notation):
C24H22N2O
CAS Number:
Molecular Weight:
354.44
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing