![(−)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%](/deepweb/assets/sigmaaldrich/product/structures/147/578/68dbd2be-b198-47d0-9420-fa4cbb574088/640/68dbd2be-b198-47d0-9420-fa4cbb574088.png)
(−)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate
![(−)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%](/deepweb/assets/sigmaaldrich/product/structures/147/578/68dbd2be-b198-47d0-9420-fa4cbb574088/640/68dbd2be-b198-47d0-9420-fa4cbb574088.png)
Synonym(s):
(−)-Diaceton-2-keto-L-gulonsäure, (−)-Diacetone-2-keto-L-gulonic acid, 2,3:4,6-Di-O-isopropyliden-α-L-xylo-2-hexulofuranosonsäure, 2,3:4,6-Di-O-isopropylidene-α-L-xylo-2-hexulofuranosonic acid, Dikegulac
Empirical Formula (Hill Notation):
C12H18O7 · H2O
CAS Number:
Molecular Weight:
292.28
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing