![1-Adamantyl isocyanide 95%](/deepweb/assets/sigmaaldrich/product/structures/276/106/21597d1e-422b-4ed8-b442-9db90fd7dee7/640/21597d1e-422b-4ed8-b442-9db90fd7dee7.png)
1-Adamantyl isocyanide
![1-Adamantyl isocyanide 95%](/deepweb/assets/sigmaaldrich/product/structures/276/106/21597d1e-422b-4ed8-b442-9db90fd7dee7/640/21597d1e-422b-4ed8-b442-9db90fd7dee7.png)
Empirical Formula (Hill Notation):
C11H15N
CAS Number:
Molecular Weight:
161.24
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing