![1-(5-Isoquinolinesulfonyl)-3-methylpiperazine ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/259/293/c8583d1d-eee5-4f5c-9bc4-ed4b0f1b3279/640/c8583d1d-eee5-4f5c-9bc4-ed4b0f1b3279.png)
1-(5-Isoquinolinesulfonyl)-3-methylpiperazine
![1-(5-Isoquinolinesulfonyl)-3-methylpiperazine ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/259/293/c8583d1d-eee5-4f5c-9bc4-ed4b0f1b3279/640/c8583d1d-eee5-4f5c-9bc4-ed4b0f1b3279.png)
Synonym(s):
1-(5-Isoquinolinylsulfonyl)-3-methylpiperazine, Iso-H-7
Empirical Formula (Hill Notation):
C14H17N3O2S
CAS Number:
Molecular Weight:
291.37
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing