Product Name
N-(2-Hydroxybenzoyl)pyrrolidine 97%
Product Number
642932
Product Brand
aldrich
CAS Number
98841-68-0
Molecular Weight
191.23
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN POWDER OR CRYSTALS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 66.7%-71.5% |
NITROGEN 7.0%- 7.6% | |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED DECEMBER 10, 2008 RJM |