Product Name
Ethyl bis(ethylthio)acetate 99%
Product Number
226343
Product Brand
aldrich
CAS Number
20461-95-4
Molecular Weight
208.34
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
IDENTITY | CONFORMS TO STRUCTURE BY IR OR HNMR SPECTRA. |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED JANUARY 29, 2008 RJM |