Skip to Content
Merck
All Photos(1)

Key Documents

SY3H7CF29147

Synthonix Corporation

5,6-Dichloro-1H-pyrrolo[3,2-b]pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
US$184.00
250 MG
US$310.50
1 G
US$632.50

Vendor SKU: D63997-100MG

US$184.00


Ships in 10 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
100 MG
US$184.00
250 MG
US$310.50
1 G
US$632.50

About This Item

CAS Number:
Molecular Weight:
187.03
MDL number:

Vendor SKU: D63997-100MG

US$184.00


Ships in 10 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

5,6-dichloro-1H-pyrrolo[3,2-b]pyridine

Inchi Code

1S/C7H4Cl2N2/c8-4-3-6-5(1-2-10-6)11-7(4)9/h1-3,10H

InChI key

CXIHYPCWVXNKAP-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

95%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/63997.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://synthonix.com/63997.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service