Recommended Products
General description
Linear peptide corresponding to human CALCRL.
Application
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected CALCRL in 10 µg of mouse brain, mouse cerebellum, mouse hypothalamus, and human pituitary tissue lysate.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected CALCRL in human lung and human heart tissue.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected CALCRL in human lung and human heart tissue.
Quality
Evaluated by Western Blotting in human lung tissue lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CALCRL in 10 µg of human lung tissue lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CALCRL in 10 µg of human lung tissue lysate.
Physical properties
~55 kDa observed
Physical form
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service