Skip to Content
Merck
All Photos(1)

Documents

552108

Millipore

Anti-Rab5 Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

General description

Recognizes the Rab5 protein.
This Anti-Rab5 Rabbit pAb is validated for use in ELISA, Immunoblotting, Immunohistochemistry for the detection of Rab5.

Immunogen

Human Retina
recombinant, human retinal Rab5A

Application

ELISA (1:4000)

Immunoblotting (1:8000)

Immunohistochemistry (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Does not cross-react with Cdc42, Rab1A, Rab3A, Rab8, Rab11B, Rab13, Rab15, Rab18, Rac1, or RhoB. Variables associated with assay conditions will dictate the proper working dilutions.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohd Sayeed et al.
The Journal of biological chemistry, 293(35), 13509-13523 (2018-07-12)
Adiponectin is a fat tissue-derived adipokine with beneficial effects against diabetes, cardiovascular diseases, and cancer. Accordingly, adiponectin-mimetic molecules possess significant pharmacological potential. Oligomeric states of adiponectin appear to determine its biological activity. We identified a highly conserved, 13-residue segment (ADP-1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service