Skip to Content
Merck
All Photos(1)

Key Documents

LIFH0483048C

Life Chemicals Inc.

3-(benzylamino)butanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
US$53.30
500 MG
US$70.20
1 G
US$89.70
2.5 G
US$210.60
5 G
US$327.60
10 G
US$556.40

Vendor SKU: F1911-3044-250MG

US$53.30


Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
250 MG
US$53.30
500 MG
US$70.20
1 G
US$89.70
2.5 G
US$210.60
5 G
US$327.60
10 G
US$556.40

About This Item

CAS Number:
Molecular Weight:
193.25
MDL number:

Vendor SKU: F1911-3044-250MG

US$53.30


Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

3-(benzylamino)butanoic acid

Inchi Code

1S/C11H15NO2/c1-9(7-11(13)14)12-8-10-5-3-2-4-6-10/h2-6,9,12H,7-8H2,1H3,(H,13,14)

InChI key

QIXZNMWEYBPTEQ-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

solid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service