Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249CE3F

J & W Pharmlab LLC

1-Piperidin-4-yl-1H-benzoimidazole dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
US$163.75
100 MG
US$203.75
250 MG
US$281.25
500 MG
US$436.25
1 G
US$747.50
5 G
US$2,497.50

Vendor SKU: 30R0052S-50MG

US$163.75


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
50 MG
US$163.75
100 MG
US$203.75
250 MG
US$281.25
500 MG
US$436.25
1 G
US$747.50
5 G
US$2,497.50

About This Item

CAS Number:
Molecular Weight:
274.19
MDL number:

Vendor SKU: 30R0052S-50MG

US$163.75


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

1-(piperidin-4-yl)-1H-benzo[d]imidazole dihydrochloride

Inchi Code

1S/C12H15N3.2ClH/c1-2-4-12-11(3-1)14-9-15(12)10-5-7-13-8-6-10;;/h1-4,9-10,13H,5-8H2;2*1H

InChI key

XOLGWORKFXYWIK-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/30R0052S

MSDS

https://www.jwpharmlab.com/home/product/30R0052S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service