Skip to Content
Merck
All Photos(1)

Key Documents

ENAH304C4266

Enamine

1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-(1H-indol-3-yl)propanoate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
463.49

Iupac Name

1,3-dioxoisoindolin-2-yl Na-(tert-butoxycarbonyl)-Na-methyltryptophanate

Inchi Code

1S/C25H25N3O6/c1-25(2,3)33-24(32)27(4)20(13-15-14-26-19-12-8-7-9-16(15)19)23(31)34-28-21(29)17-10-5-6-11-18(17)22(28)30/h5-12,14,20,26H,13H2,1-4H3

InChI key

JDFKMSBLOATWLG-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6522202

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service