Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11E1BDD3

Enamine

3,4-dibromo-5-methoxypyridine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
US$323.70
100 MG
US$483.60
250 MG
US$689.00
500 MG
US$1,085.50
1 G
US$1,392.30
2.5 G
US$2,730.00
5 G
US$4,037.80
10 G
US$5,987.80

Vendor SKU: EN300-1916094-50MG

US$323.70


Availability
Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
50 MG
US$323.70
100 MG
US$483.60
250 MG
US$689.00
500 MG
US$1,085.50
1 G
US$1,392.30
2.5 G
US$2,730.00
5 G
US$4,037.80
10 G
US$5,987.80

About This Item

CAS Number:
Molecular Weight:
266.92
MDL number:

Vendor SKU: EN300-1916094-50MG

US$323.70


Availability
Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

3,4-dibromo-5-methoxypyridine

Inchi Code

1S/C6H5Br2NO/c1-10-5-3-9-2-4(7)6(5)8/h2-3H,1H3

InChI key

SYKAIWLPBWUPCH-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1916094

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1916094


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service