Skip to Content
Merck
All Photos(1)

Key Documents

COMH039FD396

Combi-Blocks, Inc.

5-Ethoxy-3-methyl-1-phenylpyrazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
US$455.00
5 G
US$1,560.00

Vendor SKU: HI-2347-1G

US$455.00


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
1 G
US$455.00
5 G
US$1,560.00

About This Item

CAS Number:
Molecular Weight:
202.26
MDL number:

Vendor SKU: HI-2347-1G

US$455.00


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

ethyl 3-methyl-1-phenyl-1H-pyrazol-5-yl ether

Inchi Code

1S/C12H14N2O/c1-3-15-12-9-10(2)13-14(12)11-7-5-4-6-8-11/h4-9H,3H2,1-2H3

InChI key

HIXKUHSBZQQDHA-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HI-2347

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HI-2347


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service