Skip to Content
Merck
All Photos(1)

Key Documents

COM448620012

Combi-Blocks, Inc.

1-(2,4-Difluorophenyl)pyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
US$52.00
5 G
US$156.00
25 G
US$468.00

Vendor SKU: HC-3572-1G

US$52.00


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
1 G
US$52.00
5 G
US$156.00
25 G
US$468.00

About This Item

CAS Number:
Molecular Weight:
183.2
MDL number:

Vendor SKU: HC-3572-1G

US$52.00


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Iupac Name

1-(2,4-difluorophenyl)pyrrolidine

Inchi Code

1S/C10H11F2N/c11-8-3-4-10(9(12)7-8)13-5-1-2-6-13/h3-4,7H,1-2,5-6H2

InChI key

HGVQHODWFJESQW-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-3572

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-3572


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service